missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Naphthol Green B
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
It finds its applications as a leather and fur dye and as a biological stain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its applications as a leather and fur dye and as a biological stain.
Solubility
Soluble in water (160 mg/ml at 20°C), ethanol (0.9 mg/ml), and ethylene (9 mg/ml).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It finds its applications as a leather and fur dye and as a biological stain.
Solubility
Soluble in water (160 mg/ml at 20°C), ethanol (0.9 mg/ml), and ethylene (9 mg/ml).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Solubility Information | Soluble in water (160mg/ml at 20°C),ethanol (0.9mg/ml),and ethylene (9mg/ml). |
| Formula Weight | 878.47 |
| Quantity | 100 g |
| Chemical Name or Material | Naphthol Green B |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction