missing translation for 'onlineSavingsMsg'
Learn More

Narciclasine, MedChemExpress

Product Code. 16499617
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499617 5 mg 5mg
16461619 1 mg 1mg
16447078 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16499617 Supplier Medchem Express Supplier No. HY16563
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Narciclasine is a plant growth modulator. Narciclasine modulates the Rho/Rho kinase/LIM kinase/cofilin signaling pathway, greatly increasing GTPase RhoA activity as well as inducing actin stress fiber formation in a RhoA-dependent manner.

Narciclasine is a plant growth modulator. Narciclasine modulates the Rho/Rho kinase/LIM kinase/cofilin signaling pathway, greatly increasing GTPase RhoA activity as well as inducing actin stress fiber formation in a RhoA-dependent manner.

Specifications

Chemical Name or Material Narciclasine
CAS 29477-83-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C14H13NO7
Quantity 5 mg
Synonym Lycoricidinol
Solubility Information DMSO : ≥ 26 mg/mL (84.62 mM)
SMILES O=C1N[C@@]2([H])[C@H](O)[C@H](O)[C@@H](O)C=C2C3=C1C(O)=C4C(OCO4)=C3
Molecular Weight (g/mol) 307.26
Formula Weight 307.26
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.