missing translation for 'onlineSavingsMsg'
Learn More

Naringin, 97%

Product Code. 10646252 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10646252 25 g Plastic bottle 25g
10404345 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10646252 Supplier Thermo Scientific Acros Supplier No. 206910250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10236-47-2 | C27H32O14 | 580.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10236-47-2
Molecular Formula C27H32O14
Molecular Weight (g/mol) 580.54
MDL Number MFCD00148888,MFCD00149445,MFCD01461988
InChI Key DFPMSGMNTNDNHN-ZPHOTFPESA-N
PubChem CID 74787988
IUPAC Name 7-[(2S,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
SMILES C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2OC2=CC(O)=C3C(=O)C[C@H](OC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@H](O)[C@H]1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Plastic bottle
Assay Percent Range 97%
Merck Index 15, 6507
Color Beige to Yellow
Quantity 25 g
Physical Form Powder
Formula Weight 580.53
Percent Purity ≥96.0%
Chemical Name or Material Naringin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.