missing translation for 'onlineSavingsMsg'
Learn More

Naringin, 97%

Product Code. p-3756489 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10646252

Brand: Thermo Scientific Acros 206910250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10236-47-2 | C27H32O14 | 580.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10236-47-2
Molecular Formula C27H32O14
Molecular Weight (g/mol) 580.54
MDL Number MFCD00148888,MFCD00149445,MFCD01461988
InChI Key DFPMSGMNTNDNHN-ZPHOTFPESA-N
PubChem CID 74787988
IUPAC Name 7-[(2S,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
SMILES C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2OC2=CC(O)=C3C(=O)C[C@H](OC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@H](O)[C@H]1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Merck Index 15, 6507
Infrared Spectrum Authentic
Formula Weight 580.53
Percent Purity ≥96.0%
Packaging Plastic bottle
Color Beige to Yellow
Quantity 25 g
Physical Form Powder
Chemical Name or Material Naringin
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.