missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Narrow Neck Reagent Bottle
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Height (Metric):
108 mm
136 mm
170 mm
221 mm
Unit Size:
Each
Description
- HDPE, transparent
- PP tamper-evident screw cap, blue
Specifications
Specifications
| Product Type | Reagent Bottle |
| Material | HDPE |
| Includes | Polypropylene tamper-evident blue screw cap |
| Closure Material | Polypropylene |
| Capacity (Metric) | 250 mL |
| Color | Transparent |
| For Use With (Application) | Suitable for Viscous Oils |
| Height (Metric) | 136 mm |
| Length (Metric) | 60 mm |
| Width (Metric) | 60 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction