Learn More
Thermo Scientific™ National Inserts for Standard-Opening Vials
Product Code. 10688995
Click to view available options
Volume (Metric):
200 μL
225 μL
250 μL
Unit Size:
Pack of 100
Pack of 1000
Product Code. 10688995
Brand: Thermo Scientific™ C4012531L
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
With a nominal O.D. of 5mm, these Thermo Scientific™ National Inserts fit most standard-opening crimp top vials.
With a nominal O.D. of 5mm, these Thermo Scientific™ National Inserts fit most standard-opening crimp top vials.
  • Pulled point inserts are an economical choice for non-critical applications
TRUSTED_SUSTAINABILITY
Specifications
Description C4012-531L
Material Glass
Volume (Metric) 225 μL
Style Precision Point
Base Style Conical
Product Line National