missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Manual Crimpers and Decrimpers for Cleanrooms

Product Code. 15277298
Change view
Click to view available options
Size:
11 mm
13 mm
20 mm
Type:
Manual Crimper
Manual Decrimper
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Type Unit Size
15277298 13 mm Manual Crimper Each
15200379 11 mm Manual Crimper Each
15267298 11 mm Manual Decrimper Each
15210379 13 mm Manual Decrimper Each
15220379 20 mm Manual Decrimper Each
15217308 20 mm - Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15277298 Supplier Thermo Scientific™ Supplier No. C4013100SS
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Achieve secure, repeatable crimps or remove seals with minimized risk of damage.

Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use stainless-steel Thermo Scientific™ National Manual Crimpers and Decrimpers.

Crimpers provide a reproducible, secure vial closure; decrimpers allow easy removal of aluminum seals without breakage.
  • Stainless steel construction
  • Autoclavable
TRUSTED_SUSTAINABILITY

Specifications

No. per Pack 1 Ea.
Type Manual Crimper
For Use With (Equipment) Aluminum Seal
Size 13 mm
For Use With (Application) Industrial Chromatography
Product Line National

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.