missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Natural Polypropylene Gel Loading Tips
Shop All Cole-Parmer Products
Click to view available options
Volume (Metric):
1 to 200 μL
Unit Size:
Pack of 1020
Description
- 1 to 200μL
- 5 Racks of 204 tips
- Autoclavable
Specifications
Specifications
| Volume (Metric) | 1 to 200 μL |
| Color | Natural |
| Autoclavable | Autoclavable |
| Length (Metric) | 6.985cm |
| For Use With (Application) | For DNA sequencing or loading electrophoresis gels |
| Tip Style | Gel Loading |
| For Use With (Equipment) | Sartorius Biohit Proline, Costar, Eppendorf, Thermo Scientific Finnpipette, Jencons Sealpette, Oxford, Oxford Benchmate, Gilson Pipetman, Socorex, Titertek Digital |
| Material | Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction