missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Profil™ Plus 87-850 Series Yellow Natural Rubber Latex Immersion Gloves

Product Code. 12435120
Click to view available options
Size:
8
9
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 12435120

Brand: Ansell Edmont™ 87850080

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Medium weight gloves with outstanding grip. Excellent resistance to mechanical hazards and enhanced chemical resistance.100% cotton flocklining makes the glove softer and more comfortable to wear while helping to absorb perspiration. Specially treated to reduce the risk of allergy.

  • Specially treated to reduce the risk of allergy
  • Excellent resistance to mechanical hazards and enhanced chemical resistance
  • 100% cotton flocklining - soft, comfortable and absorbent
  • Ideal for janitorial use and handling in wet conditions
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN420:2003A1:2009, EN374: 2003,EN388: 2003
Coating Material Natural Rubber Latex
Color Yellow
Latex Free Yes
Material Rubber Latex
Powdered Powder Free
Product Type Gloves
Size 8
Thickness (Metric) 0.55 mm
For Use With (Application) General handling in wet conditions
Glove Cuff Style Pinked
Quantity 12 Pairs
Resistant To Mechanical hazards and enhanced chemical
Length (Metric) 300 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.