missing translation for 'onlineSavingsMsg'
Learn More

Deutsch Neumann™ Rubber Safety Pipette Fillers

Product Code. 10418882
Change view
Click to view available options
Model:
Standard 1
Universal 2
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
10418882 Universal 2 Each
10313441 Standard 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10418882 Supplier Deutsch Neumann™ Supplier No. 4700002
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reduces the risks of pipetting when handling toxic, corrosive, bacteria contaminated or bad smelling liquids. Deutsch Neumann™ Rubber Safety Pipette Fillers are manufactured from natural rubber.

  • Only two operating points
  • Excellent tensile strength and resilience
  • One-handed design is the simplest safety pipette available

Specifications

Color Red
Material Natural Rubber
Model Universal 2
Type Safety Pipette Filler
For Use With (Application) For comfortable pipetting toxic, corrosive, containing bacteria and malodorous substances
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.