missing translation for 'onlineSavingsMsg'
Learn More

Deutsch Neumann™ Rubber Safety Pipette Fillers

Reduces the risks of pipetting when handling toxic, corrosive, bacteria contaminated or bad smelling liquids. Deutsch Neumann™ Rubber Safety Pipette Fillers are manufactured from natural rubber.

5.47 € - 7.30 €

Specifications

Color Red
Material Natural Rubber
Type Safety Pipette Filler
For Use With (Application) For comfortable pipetting toxic, corrosive, containing bacteria and malodorous substances
Quantity 1 Each
View More Specs

Products 2
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
10418882
View Documents Promotion Details
Deutsch Neumann™
4700002
Universal 2
7.30 €
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
10313441
View Documents Promotion Details
Deutsch Neumann™
4700001
Standard 1
5.47 €
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
Description

Description

  • Only two operating points
  • Excellent tensile strength and resilience
  • One-handed design is the simplest safety pipette available
Specifications

Specifications

Red
Safety Pipette Filler
1 Each
Natural Rubber
For comfortable pipetting toxic, corrosive, containing bacteria and malodorous substances
Videos
SDS
Documents

Documents

Product Certifications