Learn More
Abeba™ Nature 4040 Shoes
Product Code. 15406084
Change view
Click to view available options
Size:
34
35
37
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
15406084 34 1 pair
15426084 35 1 pair
15452820 37 1 pair
15462820 39 1 pair
15472820 40 1 pair
15466084 41 1 pair
15482820 42 1 pair
15492820 43 1 pair
15486084 44 1 pair
15402830 45 1 pair
15406094 46 1 pair
15416094 47 1 pair
15436094 48 1 pair
13 options
Product Code. 15406084
Brand: Abeba™ 4040/34
Only null left
Add to basket
Add to basket
Leather covered cork footbed is ESD dissipative.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • White
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Adjustable instep area
  • Buckle fastening
  • Resistant to oil and benzine
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC
Color White
Footbed Material Cork
Material Leather
Size 34
Sole Features Slip-resistant
Sole Material Rubber
Style 4040