Learn More
Abeba™ Nature 4055 Shoes
Product Code. 15681358
Change view
Click to view available options
Size:
34
35
36
37
38
39
40
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15681358 34 1 pair
15691358 35 1 pair
15601368 36 1 pair
15611368 37 1 pair
15621368 38 1 pair
15631368 39 1 pair
15641368 40 1 pair
15651368 42 1 pair
15661368 43 1 pair
15671368 44 1 pair
15681368 45 1 pair
15691368 46 1 pair
15601378 47 1 pair
15611378 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 15681358 Supplier Abeba™ Supplier No. 4055/34
Add to basket
Add to basket
Leather covered cork footbed is ESD dissipative.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Black
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Adjustable instep area
  • Buckle fastening
  • Resistant to oil and petrol
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC
Color Black
Footbed Material Cork
Material Leather
Size 34
Sole Features Slip-resistant
Sole Material Rubber
Style 4055