Learn More
Abeba™ Nature 4085 Shoes
Product Code. 15631448
Change view
Click to view available options
Size:
34
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
15631448 40 1 pair
15671438 34 1 pair
15681438 35 1 pair
15691438 36 1 pair
15601448 37 1 pair
15611448 38 1 pair
15621448 39 1 pair
15641448 41 1 pair
15651448 42 1 pair
15661448 43 1 pair
15671448 44 1 pair
15681448 45 1 pair
15691448 46 1 pair
15601458 47 1 pair
15611458 48 1 pair
15 options
Product Code. 15631448
Brand: Abeba™ 4085/40
Only null left
Add to basket
Add to basket
Leather covered cork footbed is ESD dissipative.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Black
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • 2 adjustable instep straps
  • velcro fastening
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC
Color Black
Footbed Material Cork
Material Leather
Size 40
Sole Features Slip-resistant
Sole Material Rubber
Style 4085