Learn More
Abeba™ Nature 4086 Shoes
Product Code. 15621458
Change view
Click to view available options
Size:
34
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15621458 34 1 pair
15631458 35 1 pair
15641458 36 1 pair
15651458 37 1 pair
15661458 38 1 pair
15671458 39 1 pair
15681458 40 1 pair
15691458 41 1 pair
15601468 42 1 pair
15611468 43 1 pair
15621468 44 1 pair
15631468 45 1 pair
15641468 46 1 pair
15651468 47 1 pair
15661468 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 15621458 Supplier Abeba™ Supplier No. 4086/34
Only null left
Add to basket
Add to basket
Leather covered cork footbed is ESD dissipative.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Velours
  • Blue
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • 2 adjustable instep straps
  • Buckle fastening
  • Metal-free
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC
Color Blue
Footbed Material Cork
Material Velour
Size 34
Sole Features Slip-resistant
Sole Material Rubber
Style 4086