missing translation for 'onlineSavingsMsg'
Learn More

Tiagabine hydrochloride, MedChemExpress

Product Code. 16468578
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16468578

Brand: Medchem Express HYB0696A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tiagabine hydrochloride is a potent and selective GABA reuptake inhibitor, used as an anticonvulsant agent, with IC50s of 67, 446 and 182 nM for [3H]GABA uptake in Synaptosomes, Neurons and Glia, respectively.

Tiagabine hydrochloride is a potent and selective GABA reuptake inhibitor, used as an anticonvulsant agent, with IC50s of 67, 446 and 182 nM for [3H]GABA uptake in Synaptosomes, Neurons and Glia, respectively.

Specifications

Chemical Name or Material Tiagabine hydrochloride
CAS 145821-59-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H26ClNO2S2
Quantity 10 mg
Synonym NO050328 hydrochloride NO328 hydrochloride TGB hydrochloride
Solubility Information H2O : 100 mg/mL (242.71 mM; Need ultrasonic) ∣DMSO : ≥ 53 mg/mL (128.64 mM)
SMILES O=C([C@H]1CN(CC/C=C(C2=C(C)C=CS2)/C3=C(C)C=CS3)CCC1)O.[H]Cl
Molecular Weight (g/mol) 412.01
Formula Weight 412.01
Percent Purity 98.74%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.