missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NBC 19
GREENER_CHOICE

Product Code. 18769256
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18769256

Brand: Tocris Bioscience™ 6722/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent NLRP3 inflammasome inhibitor

NBC 19 is a potent NLRP3 inflammasome inhibitor (IC50 = 60-80 nM). Inhibits nigericin- and ATP-induced IL-1β release.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (T4)-[2-Acetyl-4,4,4-trichloro-N-cyclohexyl-3-(imino-κN)butanamidato-κO]diphenylboron
CAS 2068818-75-5
Quantity 10 mg
Target Inflammasomes Inhibitors
Molecular Formula C24H26BCl3N2O2
Formula Weight 2172
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.