missing translation for 'onlineSavingsMsg'
Learn More

NBI-98782, MedChemExpress

Product Code. 16489377
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489377 5 mg 5mg
16446838 10 mg 10mg
16442907 50 mg 50mg
16426946 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16489377 Supplier Medchem Express Supplier No. HY15793
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress NBI-98782(alpha-dihydrotetrabenazine) is a vesicular monoamine transporter (VMAT2) inhibtior with an Ki value of 0.97 nM.

NBI-98782(alpha-dihydrotetrabenazine) is a vesicular monoamine transporter (VMAT2) inhibtior with an Ki value of 0.97 nM.

Specifications

Chemical Name or Material NBI-98782
CAS 85081-18-1
Color White
Purity Grade Notes Research
Molecular Formula C19H29NO3
Quantity 5 mg
Synonym (+)-DTBZ (+)-α-Dihydrotetrabenazine (+)-α-DHTBZ
Solubility Information DMSO : 33.33 mg/mL (104.34 mM; Need ultrasonic)
SMILES COC(C=C1CCN2C[C@@H](CC(C)C)[C@@H]3O)=C(C=C1[C@]2(C3)[H])OC
Molecular Weight (g/mol) 319.44
Formula Weight 319.44
Percent Purity 98.38%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.