missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Imetit dihydrobromide

Product Code. 30002172
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30002172 50 mg 50mg
18025992 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30002172 Supplier Tocris Bioscience™ Supplier No. 0729/50
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Standard H3 and H4 agonist (H3 > H4)

An extremely potent, high affinity agonist at H3 and H4 receptors (Ki values are 0.3 and 2.7 nM respectively). Induces shape change in eosinophils with an EC50 of 25 nM. Centrally active following systemic administration.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32385-58-3
Molecular Formula C6H12Br2N4S

Specifications

Quantity 50 mg
Formula Weight 332.06
Purity >98%
Molecular Weight (g/mol) 332.06
Chemical Name or Material Imetit dihydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.