missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NBQX disodium salt
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
Description
NBQX disodium salt is a selective and competitive AMPA and kainate receptor antagonist (IC50 = 0.15 μM and 4.8 μM, respectively). NBQX blocks the antidepressant effects of 8-Hydroxy-DPAT hydrobromide (Cat. No. 0529), decreases mTOR and BDNF levels. NBQX is neuroprotective, anticonvulsant, antinociceptive and active in vivo.
NBQX disodium salt is a more water-soluble form of NBQX (Cat. No. 0373).
Specifications
Specifications
| Chemical Name or Material | 2,3-Dioxo-6-nitro-1,2,3,4-tetrahydrobenzo[f]quinoxaline-7-sulfonamide disodium salt |
| CAS | 479347-86-9 |
| Assay Percent Range | >98% |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in water |
| Molecular Formula | C12H6N4O6SNa2 |
| Formula Weight | Observed MW: 380.24 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction