missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NBQX disodium salt
GREENER_CHOICE

Product Code. 18745757
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
This item is not returnable. View return policy

Product Code. 18745757

Brand: Tocris Bioscience™ 1044/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent AMPA antagonist; more water soluble form of NBQX (Cat. No. 0373)

NBQX disodium salt is a selective and competitive AMPA and kainate receptor antagonist (IC50 = 0.15 μM and 4.8 μM, respectively). NBQX blocks the antidepressant effects of 8-Hydroxy-DPAT hydrobromide (Cat. No. 0529), decreases mTOR and BDNF levels. NBQX is neuroprotective, anticonvulsant, antinociceptive and active in vivo.

NBQX disodium salt is a more water-soluble form of NBQX (Cat. No. 0373).

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dioxo-6-nitro-1,2,3,4-tetrahydrobenzo[f]quinoxaline-7-sulfonamide disodium salt
CAS 479347-86-9
Assay Percent Range >98%
Quantity 1 mg
Solubility Soluble to 100mM in water
Molecular Formula C12H6N4O6SNa2
Formula Weight Observed MW: 380.24

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.