missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NCT 02

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
NCT 02 acts as a molecular glue that induces proteasomal degradation of CCNK and CDK12 in colorectal cancer spheroid cells in vitro (IC50 ≤ 50 μM). NCT 02 also induces apoptosis and the downregulation of genes involved in DNA damage repair.
Specifications
Specifications
| Chemical Name or Material | 6,7-Dihydro-N-(5-methyl-2-thiazolyl)-5 H-indeno[5,6-b]furan-3-acetamide |
| CAS | 790245-61-3 |
| Quantity | 10 mg |
| Target | Molecular Glues |
| Molecular Formula | C17H16N2O2S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction