missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NCT 501 hydrochloride
GREENER_CHOICE

Product Code. 18756618
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18756618

Brand: Tocris Bioscience™ 5934/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective ALDH1A1 inhibitor

NCT 501 hydrochloride is a potent and selective ALDH1A1 inhibitor (IC50 = 40 nM). Exhibits >1000-fold selectivity for ALDH1A1 over ALDH3A1, ALDH1B1, ALDH2, HPGD and HSD17β4. Also selective over a panel of 168 GPCRs. Blood-brain barrier permeable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-[[4-(Cyclopropylcarbonyl)-1-piperazinyl]methyl]-3,7-dihydro-1,3-dimethyl-7-(3-methylbutyl)-1H-purine-2,6-dione hydrochloride
CAS 1307896-46-3
Quantity 10 mg
Solubility Soluble to 100mM in water and to 100mM in DMSO
Molecular Formula C21H32N6O3.HCl
Formula Weight 452.98
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.