missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDNF Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
201.00 € - 468.00 €
Specifications
| Antigen | NDNF |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
NDNF Polyclonal antibody specifically detects NDNF in Mouse, Rat samples. It is validated for Western BlotSpecifications
| NDNF | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 79625 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| chromosome 4 open reading frame 31, fibronectin type-III domain-containing protein C4orf31, FLJ23191 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-240 of human NDNF (NP_078850.3). QKLPTRDEELFQMQIRDKAFFHDSSVIPDGAEISSYLFRDTPKRYFFVVEEDNTPLSVTVTPCDAPLEWKLSLQELPEDRSGEGSGDLEPLEQQKQQIINEEGTELFSYKGNDVEYFISSSSPSGLYQLDLLSTEKDTHFKVYATTTPESDQPYPELPYDPRVDVTSLGRTTVTLAWKPSPTASLLKQPIQYCVVINKEHNFKSLCAVEAKLSADDAFMMA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title