missing translation for 'onlineSavingsMsg'
Learn More

(-)-Nebivolol-d4 Hydrochloride, TRC

Product Code. 30473082
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473082 1 mg 1mg
30477759 10 mg -
30481451 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30473082 Supplier Toronto Research Chemicals Supplier No. TRCN3878971MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C22H22D4ClF2NO4
Molecular Weight (g/mol) 445.92
Synonym (αS,α’S,2R,2’S)-α,α’-[Iminobis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol]-d4 Hydrochloride, [2S-[2R*[R*[R*(S*)]]]]-α,α'-[Iminobis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol-d4 Hydrochloride, R-67145-d4 Hydrochloride, Levonebivolol-d4 Hydrochloride, l-Nebivolol-d4 Hydrochloride
SMILES FC1=CC(CC[C@@]([C@@H](O)C([2H])([2H])NC([2H])([2H])[C@H](O)[C@]2([H])CCC(C=C(F)C=C3)=C3O2)([H])O4)=C4C=C1.Cl

Specifications

Quantity 1 mg
Chemical Name or Material (-)-Nebivolol-d4 Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.