missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Nebivolol hydrochloride

Product Code. 18095182
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18095182 10 mg 10mg
18075792 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18095182 Supplier R&D Systems™ Supplier No. 3517/10
  • ONLINE EXCLUSIVE PRICE
    164.72 €
    148.05 € / 10mg
    Save 16.67 €
    10% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Highly selective β1-adrenoceptor antagonist

Highly selective β1-adrenoceptor antagonist (Ki values are 0.88, 20, 44, 700, 1160, 2400 and 4000 nM at β1, 5-HT1A, β2, 5-HT2, α1, H1 and D2 receptors respectively). Induces vasodilation via a nitric oxide- and cGMP-dependent mechanism (EC50 = 11.36 μM in renal arteries) and displays antihypertensive activity in vivo. GRK/β -arrestin biased agonist at the β2 adrenoceptor.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152520-56-4
Molecular Formula C22H26ClF2NO4

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 441.9
Purity >98%
Molecular Weight (g/mol) 445.924
Chemical Name or Material Nebivolol hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.