missing translation for 'onlineSavingsMsg'
Learn More

Nebivolol Hydrochloride, TRC

Product Code. 30480188
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480188 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30480188 Supplier Toronto Research Chemicals Supplier No. TRCN38793550MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nebivolol Hydrochloride
CAS 152520-56-4
Purity Grade Notes HPLC
Molecular Formula C22 H25 F2 N O4 . Cl H
Quantity 50 mg
Synonym (αR,α'R,2R,2'S)-rel-α,α'-[Iminobis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol hydrochloride (1:1), (αR,α'R,2R,2'S)-rel-α,α'-[Iminobis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol hydrochloride, [2R*[R*[R*(S*)]]]-(±)-α,α'-[Iminobis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol hydrochloride, Nebicard, Nebilet, Nebivolol hydrochloride, Nodon, R 067555
InChI Formula InChI=1S/C22H25F2NO4.ClH/c23-15-3-7-19-13(9-15)1-5-21(28-19)17(26)11-25-12-18(27)22-6-2-14-10-16(24)4-8-20(14)29-22;/h3-4,7-10,17-18,21-22,25-27H,1-2,5-6,11-12H2;1H/t17-,18-,21-,22+;/m1./s1
SMILES Cl.O[C@H](CNC[C@@H](O)[C@H]1CCc2cc(F)ccc2O1)[C@@H]3CCc4cc(F)ccc4O3
IUPAC Name (1R)-1-[(2R)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]-2-[[(2R)-2-[(2S)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]-2-hydroxyethyl]amino]ethanol;hydrochloride
Molecular Weight (g/mol) 441.9
Formula Weight 441.1518
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.