missing translation for 'onlineSavingsMsg'
Learn More

(+)-Nebivolol, TRC

Product Code. 30466589
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466589 0.5 mg 0.5mg
30472987 2.5 mg 2.5mg
30482893 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30466589 Supplier Toronto Research Chemicals Supplier No. TRCN3878960.5MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118457-15-1
Molecular Formula C22 H25 F2 N O4
Molecular Weight (g/mol) 405.44
Synonym (αR,α'R,2R,2'S)-α,α'-[Iminobis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol, 2H-1-Benzopyran-2-methanol, α,α'-[iminobis(methylene)]bis[6-fluoro-3,4-dihydro-, [2R-[2R*[R*[R*(S*)]]]]-, (+)-Nebivolol, (S,R,R,R)-Nebivolol, (αR,α'R,2R,2'S)-α,α'-[Iminobis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol], Dexnebivolol, R 67138, d-Nebivolol
IUPAC Name (1R)-1-[(2S)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]-2-[[(2R)-2-[(2R)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]-2-hydroxyethyl]amino]ethanol
SMILES O[C@H](CNC[C@@H](O)[C@H]1CCc2cc(F)ccc2O1)[C@@H]3CCc4cc(F)ccc4O3

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C22H25F2NO4/c23-15-3-7-19-13(9-15)1-5-21(28-19)17(26)11-25-12-18(27)22-6-2-14-10-16(24)4-8-20(14)29-22/h3-4,7-10,17-18,21-22,25-27H,1-2,5-6,11-12H2/t17-,18-,21-,22+/m1/s1
Formula Weight 405.1752
Chemical Name or Material (+)-Nebivolol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.