missing translation for 'onlineSavingsMsg'
Learn More

CleanSpace™ Neck Support

CleanSpace™ Neck Support is designed to be used with the CleanSpace™ Ultra and CleanSpace™ EX PAPR units.

Brand:  CleanSpace™ PAF-1013

Additional Details : Weight : 0.01200kg

 View more versions of this product

Product Code. 15930082

  • 59.35 € / Each

Estimated Shipment: 07-02-2023
to see stock.
Add to basket

Description

Description

  • Used with the revolutionary CleanSpace™Respirators: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and breathability
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement
  • Made of polyethylene plastic and is safe for use in decontaminated operations
Specifications

Specifications

AS/NZS1716: 2012, EN 12942
Neck Support
Welding, woodworking, manufacturing, smelting, construction, recycling plants, emergency services, mining, agriculture, processing plants, grinding
Store at -10°C to +55°C
Black
Large
PAF-1060, PAF-1070
8 g
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers