missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Necrostatin-1

Product Code. 18065682
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18065682 10 mg 10mg
18005482 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18065682 Supplier R&D Systems™ Supplier No. 2324/10
  • ONLINE EXCLUSIVE PRICE
    198.49 €
    178.50 € / 10mg
    Save 19.99 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. RIP1 kinase inhibitor; inhibits necroptosis

ATP-competitive, allosteric inhibitor of receptor-interacting protein kinase 1 (RIPK1) (EC50 = 182 nM). Blocks non-apoptotic cell death via inhibition of a specific cellular pathway, necroptosis, which leads to necrosis (EC50 = 494 nM). Reduces ischemic brain injury in a mouse model of stroke.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4311-88-0
Molecular Formula C13H13N3OS

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 259.33
Purity 98%
Molecular Weight (g/mol) 259.327
Chemical Name or Material Necrostatin-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.