Learn More
Necrostatin-1
Description
Necrostatin-1 functions as an allosteric inhibitor of the death domain receptor-associated adaptor kinase RIP (RIP1) in the necroptosis pathway. While affecting this distinct cell death pathway, Necrostatin-1 does not present any perturbation of the Fas/TNFR triggered canonical apoptosis cascade.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Necrostatin-1 functions as an allosteric inhibitor of the death domain receptor-associated adaptor kinase RIP (RIP1) in the necroptosis pathway. While affecting this distinct cell death pathway, Necrostatin-1 does not present any pe
Spécification
Spécification
| CAS | 4311-88-0 |
| Color | Yellow |
| Molecular Formula | C13H13N3OS |
| MDL Number | MFCD00056916 |
| Quantity | 20 mg |
| Sensitivity | Light sensitive |
| Synonym | necrostatin-1, mth-dl-tryptophan, nec-1, necrostatin 1, 5-1h-indol-3-ylmethyl-3-methyl-2-thioxo-4-imidazolidinone, mth-trp, methyl-thiohydantoin-tryptophan, necroptotic inhibitor, nec-1, 5-1h-indol-3-ylmethyl-3-methyl-2-sulfanylideneimidazolidin-4-one, 4-imidazolidinone, 5-1h-indol-3-ylmethyl-3-methyl-2-thioxo |
| Solubility Information | Soluble in DMSO (10-25mg/ml),100% ethanol (6mg/ml),and methanol (5mg/ml). |
| InChI Key | TXUWMXQFNYDOEZ-UHFFFAOYSA-N |
| SMILES | CN1C(=O)C(NC1=S)CC2=CNC3=CC=CC=C32 |
| Afficher plus |
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.