missing translation for 'onlineSavingsMsg'
Learn More

Necrostatin-1, TRC

Product Code. 30483890
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30483890 50 mg
30476524 10 mg
30480354 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30483890 Supplier Toronto Research Chemicals Supplier No. TRCN38855050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4311-88-0
Molecular Formula C13 H13 N3 O S
Molecular Weight (g/mol) 259.33
Synonym 5-(1H-Indol-3-ylmethyl)-3-methyl-2-thioxo-4-imidazolidinone
IUPAC Name 5-(1H-indol-3-ylmethyl)-3-methyl-2-sulfanylideneimidazolidin-4-one
SMILES CN1C(=S)NC(Cc2c[nH]c3ccccc23)C1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H13N3OS/c1-16-12(17)11(15-13(16)18)6-8-7-14-10-5-3-2-4-9(8)10/h2-5,7,11,14H,6H2,1H3,(H,15,18)
Formula Weight 259.0779
Chemical Name or Material Necrostatin-1
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.