Learn More
Trajan Scientific and Medical™ Needle cleaning kit includes stylet wires and tweezers SGE
Product Code. 10422561
Change view
Click to view available options
Includes:
Tube of Stylet Wires 150mm x 0.10mm O.D., Tube of Stylet Wires 150mm x 0.15mm O.D., Tube of Stylet Wires 150mm x 0.20mm O.D., 20mL Cleaning Solution Concentrate, Tweezer
Unit Size:
Each
Product Code. Includes unitSize
10422561 Tube of Stylet Wires 150mm x 0.10mm O.D., Tube of Stylet Wires 150mm x 0.15mm O.D., Tube of Stylet Wires 150mm x 0.20mm O.D., 20mL Cleaning Solution Concentrate, Tweezer Each
1 options
Product Code. 10422561 Supplier Trajan Scientific and Medical™ Supplier No. 031782
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
SGE syringe stylet wires are designed for effective cleaning of syringe needles
  • Effective cleaning of syringe needle
  • Kit available as well as replacement wires
Specifications
Includes Tube of Stylet Wires 150mm x 0.10mm O.D., Tube of Stylet Wires 150mm x 0.15mm O.D., Tube of Stylet Wires 150mm x 0.20mm O.D., 20mL Cleaning Solution Concentrate, Tweezer
Product Type Needle Cleaning Kit