missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Needle-Valve Stopcock and Burette Lateral Stopcock
Stopcock with interchangeable PTFE valve spindle, a cap, and a retention device.
44.95 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10334901
|
BRAND™
82120 |
2 to 10 mL Burette |
44.95 €
Each |
In Stock
Log in to see stock. |
|||||
10604461
|
BRAND™
82121 |
25 mL Burette |
44.95 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
10048090
|
BRAND™
82122 |
50 mL Burette |
44.95 €
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
- Valve housing with precision Borosilicate Glass 3.3 burette tip
- Interchangeable PTFE valve spindle, PP cap, and retention device
- Valve opening 0 to 2.5 mm
Specifications
Stopcock | |
0 to 2.5 mm |