missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Needle-Valve Stopcock and Burette Lateral Stopcock

Product Code. 10048090 Shop All Brand Products
Click to view available options
For Use With (Equipment):
2 to 10 mL Burette
25 mL Burette
50 mL Burette
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10048090

Brand: BRAND™ 82122

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Stopcock with interchangeable PTFE valve spindle, a cap, and a retention device.

  • Valve housing with precision Borosilicate Glass 3.3 burette tip
  • Interchangeable PTFE valve spindle, PP cap, and retention device
  • Valve opening 0 to 2.5 mm
TRUSTED_SUSTAINABILITY

Specifications

Type Stopcock
Diameter (Metric) 0 to 2.5 mm
For Use With (Equipment) 50 mL Burette
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.