Learn More
BRAND™ Needle-Valve Stopcock and Spare Spindle
Product Code. 10605941 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric):
0 to 2.5 mm
Unit Size:
Each
Product Code. Diameter (Metric) unitSize
10605941 0 to 2.5 mm Each
1 options
Product Code. 10605941 Supplier BRAND™ Supplier No. 82170
In Stock
Add to basket
Add to basket
Needle-Valve Stopcock and Spare Spindle constructed of PTFE with retention device.
  • Interchangeable
  • PTFE valve spindle
  • PP cap and retention device
  • Valve opening 0 to 2.5 mm
TRUSTED_SUSTAINABILITY
Specifications
Type Valve Spindle
Diameter (Metric) 0 to 2.5 mm
Material PTFE
Includes Retention Device