missing translation for 'onlineSavingsMsg'
Learn More

Nefazodone hydrochloride, 98+%

Product Code. 15425099
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425099 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15425099 Supplier Thermo Scientific Alfa Aesar Supplier No. J62793.03
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 82752-99-6 | C25H33Cl2N5O2 | 506.47 g/mol

An antidepressant acts by modifying serotonin transmission. Mixed 5-HT2A serotonin receptor antagonist/serotonin uptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An antidepressant acts by modifying serotonin transmission
  • Mixed 5-HT2A serotonin receptor antagonist/serotonin uptake inhibitor
TRUSTED_SUSTAINABILITY

Specifications

CAS 82752-99-6
Melting Point 186°C to 187°C
Color White
Molecular Formula C25H33Cl2N5O2
MDL Number MFCD00935760
Quantity 1 g
Merck Index 14,6438
Synonym nefazodone hydrochloride, nefazodone hcl, serzone, dutonin, menfazona, rulivan, reseril, 1-3-4-3-chlorophenyl piperazin-1-yl propyl-3-ethyl-4-2-phenoxyethyl-1h-1,2,4-triazol-5 4h-one hydrochloride, nefadar
InChI Key DYCKFEBIOUQECE-UHFFFAOYSA-N
SMILES [H+].[Cl-].CCC1=NN(CCCN2CCN(CC2)C2=CC=CC(Cl)=C2)C(=O)N1CCOC1=CC=CC=C1
IUPAC Name hydrogen 1-{3-[4-(3-chlorophenyl)piperazin-1-yl]propyl}-3-ethyl-4-(2-phenoxyethyl)-4,5-dihydro-1H-1,2,4-triazol-5-one chloride
Molecular Weight (g/mol) 506.47
PubChem CID 54911
ChEBI CHEBI:7495
Formula Weight 506.47
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Nefazodone hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.