missing translation for 'onlineSavingsMsg'
Learn More

Nefiracetam, MedChemExpress

Product Code. 16468246
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468246 100 mg 100mg
16404337 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468246 Supplier Medchem Express Supplier No. HYB0340
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nefiracetam is a GABAergic, cholinergic, and monoaminergic neuronal systems enhancer for Ro 5-4864-induced convulsions.

Nefiracetam is a GABAergic, cholinergic, and monoaminergic neuronal systems enhancer for Ro 5-4864-induced convulsions.

Specifications

Chemical Name or Material Nefiracetam
CAS 77191-36-7
Color White
Purity Grade Notes Research
Molecular Formula C14H18N2O2
Quantity 100 mg
Synonym DM9384 DZL-221
Solubility Information DMSO : ≥ 100 mg/mL (406.01 mM)
SMILES O=C(NC1=C(C)C=CC=C1C)CN2C(CCC2)=O
Molecular Weight (g/mol) 246.3
Formula Weight 246.3
Percent Purity 99.39%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.