missing translation for 'onlineSavingsMsg'
Learn More

Nelfinavir Hydroxy-tert-butylamide, TRC

Product Code. 30476828
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476828 1 mg 1mg
30467248 10 mg -
30470292 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30476828 Supplier Toronto Research Chemicals Supplier No. TRCN3897601MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213135-56-9
Molecular Formula C32 H45 N3 O5 S
Molecular Weight (g/mol) 583.78
IUPAC Name (3S,4aS,8aS)-2-[(2R,3R)-2-hydroxy-3-[(3-hydroxy-2-methylbenzoyl)amino]-4-phenylsulfanylbutyl]-N-(1-hydroxy-2-methylpropan-2-yl)-3,4,4a,5,6,7,8,8a-octahydro-1H-isoquinoline-3-carboxamide
SMILES Cc1c(O)cccc1C(=O)N[C@@H](CSc2ccccc2)[C@H](O)CN3C[C@H]4CCCC[C@H]4C[C@H]3C(=O)NC(C)(C)CO

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C32H45N3O5S/c1-21-25(14-9-15-28(21)37)30(39)33-26(19-41-24-12-5-4-6-13-24)29(38)18-35-17-23-11-8-7-10-22(23)16-27(35)31(40)34-32(2,3)20-36/h4-6,9,12-15,22-23,26-27,29,36-38H,7-8,10-11,16-20H2,1-3H3,(H,33,39)(H,34,40)/t22-,23+,26-,27-,29+/m0/s1
Formula Weight 583.308
Chemical Name or Material Nelfinavir hydroxy-tert-butylamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.