missing translation for 'onlineSavingsMsg'
Learn More

Nemadectin, TRC

Product Code. 30477776
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477776 50 mg 50mg
30482154 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477776 Supplier Toronto Research Chemicals Supplier No. TRCN38979050MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nemadectin
CAS 102130-84-7
Molecular Formula C36 H52 O8
Quantity 50 mg
Synonym Nemadectin, Milbemycin B','5-O-demethyl-28-deoxy-25-(1,3-dimethyl-1-butenyl)-6,28-epoxy-23-hydroxy-','[6R,23S,25S(E)]- (9CI), (6R,23S)-5-O-Demethyl-28-deoxy-25-[(1E)-1,3-dimethyl-1-buten-1-yl]-6,28-epoxy-23-hydroxymilbemycin B, Spiro[11,15-methano-2H,13H,17H-furo[4,3,2
InChI Formula InChI=1S/C36H52O8/c1-20(2)13-24(6)32-25(7)30(37)18-35(44-32)17-28-16-27(43-35)12-11-22(4)14-21(3)9-8-10-26-19-41-33-31(38)23(5)15-29(34(39)42-28)36(26,33)40/h8-11,13,15,20-21,25,27-33,37-38,40H,12,14,16-19H2,1-7H3/b9-8+,22-11+,24-13+,26-10+/t21-,25-,27+,2
SMILES CC(C)\C=C(/C)\[C@H]1O[C@@]2(C[C@@H]3C[C@@H](C\C=C(/C)\C[C@@H](C)\C=C\C=C\4/CO[C@@H]5[C@H](O)C(=C[C@@H](C(=O)O3)[C@]45O)C)O2)C[C@H](O)[C@@H]1C
Molecular Weight (g/mol) 612.793
Formula Weight 612.366
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.