missing translation for 'onlineSavingsMsg'
Learn More

Neocuproine Hydrochloride, ≥98% (Dry Basis)

Product Code. 15895318 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15895318 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15895318 Supplier Thermo Scientific Supplier No. J19430.A1
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7296-20-0 | C14H13ClN2 | 244.722 g/mol

 

TRUSTED_SUSTAINABILITY

Specifications

CAS 7296-20-0
Packaging Plastic bottle
Molecular Formula C14H13ClN2
MDL Number MFCD00150062
Quantity 1 kg
Synonym neocuproine hydrochloride, 1,10-phenanthroline, 2,9-dimethyl-, hydrochloride, 1,10-phenanthroline, 2,9-dimethyl-, monohydrochloride, 2,9-dimethyl-1,10-phenanthroline hydrochloride, 2,9-dimethyl-1,10-phenanthroline hcl, 1,10-phenanthroline, 2,9-dimethyl-, hydrochloride 1:?, 1,10-phenanthroline, 2,9-dimethyl-, hydrochloride 1:1, 2,9-dimethylpyridino 3,2-h quinoline, chloride, c14h12n2.clh, neocuproine hcl
InChI Key QUGBNSJLQDNNPK-UHFFFAOYSA-N
SMILES CC1=NC2=C(C=C1)C=CC3=C2N=C(C=C3)C.Cl
IUPAC Name 2,9-dimethyl-1,10-phenanthroline;hydrochloride
Molecular Weight (g/mol) 244.722
PubChem CID 81718
Formula Weight 244.72 (Anhydrous)
Percent Purity ≥98%
Physical Form Powder
Assay Dry basis
Chemical Name or Material Neocuproine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.