missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher 80

Product Code. 10364563
Change view
Click to view available options
Volume (Metric):
10 kg
Unit Size:
10kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10364563 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10364563 Supplier Dr.Weigert™ Supplier No. 320376
  • ONLINE EXCLUSIVE PRICE
    116.00 €
    104.72 € / 10kg
    Save 11.28 €
    9% Off
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neodisher 80 - Alkaline intensive detergent powder for automated dishwashing. Removes dried on food residues and stubborn starch, tea and coffee residues.

  • Alkaline detergent, Powder
  • neodisher 80 is specially suitable for commercial fresh water and single-tank dishwashers in the gastronomy sector, in hotels, homes, ward kitchens and hospitals
  • Reliably removes dried on food residues coffee and tea residues
  • Suitable for items made of china/porcelain, stainless steel and alkali-compatible plastic
  • Not suitable for aluminium, anodized aluminium and light metal alloys
  • Dosage of 2-4 g/L
  • Usable with all degrees of water hardness

Specifications

Product Type Neodisher 80
Form Powder
pH 12.6 to 13.2
Sterility Non-sterile
Composition <5 % bleaching agents based on chlorine, aliphatic hydrocarbons > 30 % phosphates
Concentration 2 to 4 g/L
Density 1,125 to 1,150 g/L
Description 280 x 270 mm
Volume (Metric) 10 kg
compliance-icons compliance-icons
Product Identifier
  • neodisher 80
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: disodium metasilicate pentahydrate, sodium carbonate, sodiumhydroxide, troclosene sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.