missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher A8

Product Code. 10160591
Change view
Click to view available options
Volume (Metric):
25 kg
Unit Size:
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10160591 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10160591 Supplier Dr.Weigert™ Supplier No. 410266
  • ONLINE EXCLUSIVE PRICE
    262.00 €
    251.91 € / 25kg
    Save 10.09 €
    3% Off
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neodisher LaboClean A8 - Powder Detergent alkaline cleaning is to be used for cleaning and washing glassware. Applications include pathology, parasitology, dairy and food industries.

  • Alkaline detergent In powder form
  • neodisher LaboClean A8 is used for cleaning in washer disinfection
  • chlorine-based bleaching agents and phosphates, Free of surfactants
  • suitable for residue-free cleaning of tissue cultures and cell layersDosage of between 2-5 g/L

    Specifications

    Product Type Neodisher A8
    Form Powder
    pH 12 to 12.4
    Sterility Non-sterile
    Composition <5 % chlorine-based bleaching agents 15 - 30 % phosphates
    Concentration 2 to 5 g/L
    Density 1200 g/L
    Description 600 x 300 x 150 mm
    Volume (Metric) 25 kg
    compliance-icons compliance-icons compliance-icons
    Product Identifier
    • neodisher LaboClean A 8
    Signal Word
    • Danger
    Hazard Category
    • Substances and mixtures corrosive to metals Category 1
    • Skin corrosion/irritation Category 1 A
    • Serious eye damage/eye irritation Category 1
    • Specific target organ toxicity Category 3
    • LONG-TERM AQUATIC HAZARD Chronic 2
    Hazard Statement
    • H290-May be corrosive to metals.
    • H314-Causes severe skin burns and eye damage.
    • H335-May cause respiratory irritation.
    • H411-Toxic to aquatic life with long lasting effects.
    • EUH031-Contact with acids liberates toxic gas.
    Precautionary Statement
    • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
    • P273-Avoid release to the environment.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
    • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P310-Immediately call a POISON CENTER/doctor/.
    Supplemental information
    • MIXTURE LIST-Contain: disodium metasilicate pentahydrate, sodium carbonate, sodiumhydroxide, troclosene sodium
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.