missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher LaboClean A 8

Product Code. 11713733
Change view
Click to view available options
Volume (Metric):
1 kg
10 kg
Unit Size:
10kg
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11713733 10 kg 10kg
11717105 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11713733 Supplier Dr.Weigert™ Supplier No. 410276
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neodisher LaboClean A8 - Powder Detergent alkaline cleaning is to be used for cleaning and washing glassware. Applications include pathology, parasitology, dairy and food industries.

  • Alkaline detergent In powder form
  • neodisher LaboClean A 8 is used for cleaning in washer disinfection
  • chlorine-based bleaching agents and phosphates, Free of surfactants
  • suitable for residue-free cleaning of tissue cultures and cell layersDosage of between 2-5 g/L

    Spezifikation

    Product Type Neodisher A8
    Form Powder
    pH 12 to 12.4
    Sterility Non-sterile
    Composition Disodium metasilicate pentahydrate; sodium hydroxide; troclosene sodium; Sodium carbonate
    Concentration 2 to 5 g/L
    Container Type Bucket
    Description 280 x 270 mm
    For Use With (Application) Removes Blood, Serum, Food Residues, Pharmaceuticals, Paints, Varnishes, Gelatin and Other Organic Residues
    Volume (Metric) 10 kg
    compliance-icons compliance-icons compliance-icons
    Produktidentifikator
    • neodisher LaboClean A 8
    Signalwort
    • Danger
    Gefahrenkategorie
    • Substances and mixtures corrosive to metals Category 1
    • Skin corrosion/irritation Category 1 A
    • Serious eye damage/eye irritation Category 1
    • Specific target organ toxicity Category 3
    • LONG-TERM AQUATIC HAZARD Chronic 2
    Gefahrenhinweis
    • H290-May be corrosive to metals.
    • H314-Causes severe skin burns and eye damage.
    • H335-May cause respiratory irritation.
    • H411-Toxic to aquatic life with long lasting effects.
    • EUH031-Contact with acids liberates toxic gas.
    Sicherheitshinweis
    • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
    • P273-Avoid release to the environment.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
    • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P310-Immediately call a POISON CENTER/doctor/.
    Ergänzende Informationen
    • MIXTURE LIST-Contain: disodium metasilicate pentahydrate, sodium carbonate, sodiumhydroxide, troclosene sodium
    Name des Produkts
    Wählen Sie ein Thema

    Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.