missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher Brilliant Clean

Product Code. 10681324
Change view
Click to view available options
Volume (Metric):
15 kg
Unit Size:
15kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10681324 15 kg 15kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10681324 Supplier Dr.Weigert™ Supplier No. 350042
  • ONLINE EXCLUSIVE PRICE
    117.00 €
    98.15 € / 15kg
    Save 18.85 €
    16% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neodisher brilliant clean - Universal detergent powder for automated dishwashing. Especially suitable for freewater dishwasher, household dishwasher and small single-tank dishwasher.

  • Alkaline detergent, Powder
  • neodisher brilliant clean is used for automated cleaning of glasses,chinaware, cutley and working utensils in all food processing companies such as professional kitchens, butcher’s shops and bakeries
  • Offers increased hygiene safety
  • Excellent bleaching effect
  • Removes especially protein, coffee and tea residues
  • Excellent material protection
  • Dosage of 2-4 g/L
  • Usable with all degrees of water hardness

Specifications

Product Type Neodisher Brilliant Cleaner
Form Powder
pH 11.5 to 11.7
Sterility Non-sterile
Composition <5% chlorine-based bleaching agent, >30% phosphates
Concentration 2 to 4 g/L
Density 1050 g/L
Description 280 x 120 x 110 mm
Volume (Metric) 15 kg
compliance-icons compliance-icons
Product Identifier
  • neodisher brilliant clean
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: disodium metasilicate pentahydrate, sodium carbonate, troclosene sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.