missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher EM

Product Code. 11813242
Change view
Click to view available options
Volume (Metric):
5 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11813242 5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11813242 Supplier Dr.Weigert™ Supplier No. 430033
  • ONLINE EXCLUSIVE PRICE
    137.00 €
    114.85 € / Pack of 1
    Save 22.15 €
    16% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neodisher EM - Additive liquid for Improving the cleaning performance of alkaline and acidic cleaners. cleaning Additive is especially useful in removing solids, paraffin, oils and fats in aqueous solutions.

  • Additive liquid with emulsifying, dispersing and defoaming effect
  • Neodisher EM is used to improve the performance of an alkaline or acidic detergent
  • Suitable for glass, stainless steel and aluminum
  • Applications include the cosmetics industry, research, pharmaceutical industry and chemical industries
  • Dosage of between 1-6 mL/L, depending on the characteristique of the soil and degree of contamination

Specifications

Product Type Neodisher EM
Form Liquid
pH 6 to 8
Sterility Non-sterile
Composition Non-ionic surfactants
Concentration 1 to 6 mL/L
Container Type Canister
Density 0.9 g/cm3
Description 220 x 240 x 140 mm
Filtered Filtered
Volume (Metric) 5 L
Show More Show Less
compliance-icons
Product Identifier
  • neodisher EM
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: non ionic surfactants
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.