missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher FA

Product Code. 10767761
Change view
Click to view available options
Container Type:
Canister
Cannister
Unit Size:
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Container Type unitSize
10767761 Canister 5 L
11898990 Cannister 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10767761 Supplier Dr.Weigert™ Supplier No. 410133
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Neodisher LaboClean FA - Alkaline detergent, liquid concentrate for the automated cleaning of surgical thermo-stable and thermolabile instruments and others medicals or laboratories utensils. Versatile and concentrated detergent.

  • Alkaline detergent, liquid concentrate
  • neodisher LaboClean FA is used for cleaning in washer disinfection
  • Free of surfactants and active chlorine
  • Excellent material compatibility
  • Cleans blood, protein, culture media and gelatine
  • Dosage between 2-4 mL/l

Specifications

Product Type Neodisher FA
Form Liquid
pH 11.6 to 11.8
Sterility Non-sterile
Composition disodium metasilicate, potassium silicate, potassium hydoxide
Concentration 1 to 4 mL/L
Container Type Canister
Density 1.5 g/cm3 (20°C)
Filtered Filtered
Volume (Metric) 5 L
compliance-icons
Product Identifier
  • neodisher FA
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: disodium metasilicate, dipotassium metasilicate, potassium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.