missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher LaboClean FLA

Product Code. 11768373
Change view
Click to view available options
Volume (Metric):
5 L
10 L
Unit Size:
10 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11768373 5 L 5 L
11758373 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11768373 Supplier Dr.Weigert™ Supplier No. 411233
  • ONLINE EXCLUSIVE PRICE
    85.00 €
    55.67 € / 5 L
    Save 29.33 €
    34% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neodisher LaboClean FLA - Alkaline detergent, liquid concentrate for the automated cleaning of commonly used laboratory utensils (in medical, biological, microbiological, chemical, food, pharmaceutical, ... ). Removes stubborn oily residues and other organic residues, e.g. fats, oils, starch, residues of felt-tip pens

  • Alkaline detergent, liquid concentrate
  • neodisher LaboClean FLA is used for cleaning in washer disinfection
  • Free of phosphates, surfactants and active chlorine
  • Removes organic residues, especially suitable for cleaning animal cages for mice or rats made of polysulfone, polycarbonate, polyetherimide, polyphenylsulfone or stainless steel
  • Dosage of between 2-6 mL/L

Specifications

Product Type Neodisher FLA
Form Liquid
pH 12.3 to 12.8
Sterility Non-sterile
Container Type Canister
Description 220 x 240 x 140 mm
Filtered Filtered
For Use With (Application) Removes Stubborn Oily Residues and Other Organic Residues
Volume (Metric) 5 L
compliance-icons compliance-icons
Product Identifier
  • neodisher LaboClean FLA
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: potassium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.