missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher LaboClean GK

Product Code. 11723733
Change view
Click to view available options
Volume (Metric):
10 kg
Unit Size:
10kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11723733 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11723733 Supplier Dr.Weigert™ Supplier No. 410376
  • ONLINE EXCLUSIVE PRICE
    176.87 €
    157.42 € / 10kg
    Save 19.45 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Neodisher laboClean GK - Mild alkaline powder for mechanical cleaning of laboratory glassware in microbiological, virological and isotope laboratories as well as nuclear medicine laboratories and pharmacies.

  • Alkaline detergent, Powder
  • neodisher LaboClean GK is used for cleaning laboratory glassware in washer disinfection
  • Free of surfactants
  • Removes residues of nutrient media, cell cultures and tissues, radioactive contaminants and organic dye and pigment impurities
  • Dosage of 2-5 g/L
  • Easy to rinse off
  • Suitable for utensils made of glass, ceramics, quartz and plastics

Specifications

Product Type Neodisher GK
Form Powder
pH 10.8 to 11.9
Sterility Non-sterile
Composition Chlorine-based bleaching agents, phosphates
Concentration 1 to 4 g/L
Container Type Bucket
Density 915 to 965 g/L
Description 280 x 270 mm
For Use With (Application) Removes Residues of Culture Media, Residues of Cell and Tissue Cultures, Radioactive Contamination, Organic Colour and Pigment Contamination. Easy to Rinse Off
Volume (Metric) 10 kg
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • neodisher LaboClean GK
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap..
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: trisodium phosphate,troclosene sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.