missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher LaboClean LA

Product Code. 11845291
Change view
Click to view available options
Volume (Metric):
3 kg
Unit Size:
3kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11845291 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11845291 Supplier Dr.Weigert™ Supplier No. 410681
  • ONLINE EXCLUSIVE PRICE
    48.20 €
    41.23 € / 3kg
    Save 6.97 €
    14% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Neodisher LaboClean LA - Alkaline cleaning agent in powder form for use in special washing machines. For cleaning laboratory glassware and utensils for laboratories in the foodstuffs processing industry, the cosmetics, pharma, mineral industries and others laboratories. Removes oils, fats, resins, paraffin and organic dye stuffs.

  • Alkaline detergent, Powder
  • neodisher LaboClean LA is used for cleaning laboratory glassware
  • Polycarboxylates, phosphates and non-ionic surfactants
  • Suitable for utensils made of glass, ceramics, quartz and plastics
  • Dosage of 2-5 g/L
  • Contains high proportions of emulsifiers and dispersing agents
  • Exhibits a defoaming action
  • For laboratory glassware and utensils for laboratories in the foodstuffs processing industry, the cosmetics, pharma and mineral industries and other laboratories

Specifications

Product Type Neodisher LA
Form Powder
Sterility Non-sterile
Composition Polycarboxylates, non-ionic surfactants, phosphates
Concentration 2 to 5 g/L
Container Type Bucket
Description 220 x 140 mm
For Use With (Application) Suitable for The Removal of Greasy and Oily Dirt, Resins, and Paraffin
Packaging 4/Pk.
Quantity 4/Pk.
Volume (Metric) 3 kg
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • neodisher LaboClean LA
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: fatty alcohol, ethoxylated, disodium metasilicate pentahydrate, sodium carbonate, sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.